LIF945291910 Life Chemicals Inc.

2-[(6-chloropyridin-3-yl)formamido]acetic acid

  • MDL number MFCD11646710
  • Cas Number : 1099143-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart