LIF945291913 Life Chemicals Inc.

3-chloro-4-oxopentyl acetate

  • MDL number MFCD00068821
  • Cas Number : 13051-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart