LIF945291916 Life Chemicals Inc.

N-(2-aminoethyl)-3-chlorobenzene-1-sulfonamide hydrochloride

  • MDL number MFCD09971594
  • Cas Number : 1170276-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart