LIF945291920 Life Chemicals Inc.

2-(methylamino)-N-phenylacetamide hydrochloride

  • MDL number MFCD10686932
  • Cas Number : 60565-45-9
  • Molecular Weight : 200.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart