LIF945291928 Life Chemicals Inc.

4-methyl-2-(pyrrolidin-2-yl)-1,3-oxazole dihydrochloride

  • MDL number MFCD30738832
  • Cas Number : 2097936-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart