LIF945291929 Life Chemicals Inc.

5-bromo-6-chloro-4-methylpyridine-3-carbonitrile

  • MDL number MFCD29913698
  • Cas Number : 2090170-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart