LIF945291937 Life Chemicals Inc.

2-oxo-3,4-dihydro-2H-1,3-benzoxazine-6-sulfonyl chloride

  • MDL number MFCD23144046
  • Cas Number : 1427379-39-2
  • Molecular Weight : 247.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart