LIF945291938 Life Chemicals Inc.

1,3-oxazole-5-carbonitrile

  • MDL number MFCD04114936
  • Cas Number : 68776-61-4
  • Molecular Weight : 94.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart