LIF945291944 Life Chemicals Inc.

tert-butyl 4-iodo-1H-pyrazole-1-carboxylate

  • MDL number MFCD05663855
  • Cas Number : 121669-70-3
  • Molecular Weight : 294.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart