LIF945291948 Life Chemicals Inc.

N1-methyl-4-(trifluoromethyl)benzene-1,2-diamine

  • MDL number MFCD01571385
  • Cas Number : 35203-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart