LIF945291955 Life Chemicals Inc.

6-ethyl-N-(propan-2-yl)pyrimidin-4-amine

  • MDL number MFCD14603029
  • Cas Number : 1250911-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart