LIF945291973 Life Chemicals Inc.

N-[3-(aminomethyl)pyridin-2-yl]-N-methylmethanesulfonamide

  • MDL number MFCD15071408
  • Cas Number : 939791-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart