LIF945292014 Life Chemicals Inc.

6-fluoro-3-methoxy-2-methylquinolin-4-amine

  • MDL number MFCD30737463
  • Cas Number : 2095410-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart