LIF945292036 Life Chemicals Inc.

(2-ethoxy-2,2-difluoroethyl)(methyl)amine hydrochloride

  • MDL number MFCD30741149
  • Cas Number : 2097859-99-7
  • Molecular Weight : 175.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart