LIF945292048 Life Chemicals Inc.

2-chloro-6-fluoro-1,3-benzoxazole

  • MDL number MFCD16250625
  • Cas Number : 153403-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart