LIF945292052 Life Chemicals Inc.

7-phenyl-1,4-thiazepane hydrochloride

  • MDL number MFCD28895828
  • Cas Number : 2097937-23-8
  • Molecular Weight : 229.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-phenyl-1,4-thiazepane hydrochloride
Inchi Code 1S/C11H15NS.ClH/c1-2-4-10(5-3-1)11-6-7-12-8-9-13-11;/h1-5,11-12H,6-9H2;1H
Inchi Key KQHZSRBDCUJBLA-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart