LIF945292060 Life Chemicals Inc.

3-(4-chlorophenyl)morpholine hydrochloride

  • MDL number MFCD03840089
  • Cas Number : 1170797-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart