LIF945292069 Life Chemicals Inc.

1,2-oxazole-4-carbaldehyde

  • MDL number MFCD02179515
  • Cas Number : 65373-53-7
  • Molecular Weight : 97.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart