LIF945292072 Life Chemicals Inc.

4-hydroxy-3,6-dimethyl-1,2-dihydropyridin-2-one

  • MDL number MFCD11113422
  • Cas Number : 4664-13-5
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart