LIF945292076 Life Chemicals Inc.

ethyl [(3-chloro-4-methylphenyl)carbamoyl]formate

  • MDL number MFCD01077273
  • Cas Number : 349119-38-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart