LIF945292090 Life Chemicals Inc.

ethyl 5-cyclopropyl-1,2-oxazole-4-carboxylate

  • MDL number MFCD18299860
  • Cas Number : 124845-03-0
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart