LIF945292095 Life Chemicals Inc.

ethyl 5-chloro-3-phenyl-1H-indole-2-carboxylate

  • MDL number MFCD00411467
  • Cas Number : 21139-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart