LIF945292097 Life Chemicals Inc.

10-bromo-3-(2-hydroxyethyl)-3-azatricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11-pentaene-2,4-dione

  • MDL number MFCD00476635
  • Cas Number : 52559-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart