LIF945292101 Life Chemicals Inc.

5-(4-chlorophenyl)-3,4-dihydro-2H-pyrrole

  • MDL number MFCD02931038
  • Cas Number : 22217-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart