LIF945292114 Life Chemicals Inc.

N-(piperidin-4-yl)pyrimidin-4-amine

  • MDL number MFCD14605698
  • Cas Number : 1211585-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart