LIF945292120 Life Chemicals Inc.

non-8-enal

  • MDL number MFCD11553618
  • Cas Number : 39770-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart