LIF945292124 Life Chemicals Inc.

1-bromo-2-methyl-4-(propan-2-yloxy)benzene

  • MDL number MFCD18917791
  • Cas Number : 86786-24-5
  • Molecular Weight : 229.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart