LIF945292130 Life Chemicals Inc.

[3-(benzyloxy)-1-(hydroxymethyl)cyclobutyl]methanol

  • MDL number MFCD30741152
  • Cas Number : 20061-24-9
  • Molecular Weight : 222.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart