LIF947290446 Life Chemicals Inc.

1-(1-methylpiperidin-4-yl)ethan-1-amine

  • MDL number MFCD09864381
  • Cas Number : 1095499-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart