LIF947290481 Life Chemicals Inc.

(7R,8aS)-2-benzyl-7-hydroxy-octahydropyrrolo[1,2-a]piperazine-1,4-dione

  • MDL number MFCD28132965
  • Cas Number : 132714-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart