LIF947290486 Life Chemicals Inc.

2-(oxolan-2-yl)acetic acid

  • MDL number MFCD03990566
  • Cas Number : 2434-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart