LIF947290492 Life Chemicals Inc.

2-cyclohexyl-octahydropyrrolo[3,4-c]pyrrole

  • MDL number MFCD20547694
  • Cas Number : 86732-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart