LIF947290494 Life Chemicals Inc.

methyl 2-(chlorosulfonyl)cyclohex-1-ene-1-carboxylate

  • MDL number MFCD19381873
  • Cas Number : 161809-17-2
  • Molecular Weight : 238.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart