LIF947290496 Life Chemicals Inc.

2,2,2-trifluoro-N-(2-hydroxycyclohexyl)acetamide

  • MDL number MFCD00465464
  • Cas Number : 496941-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart