LIF947290497 Life Chemicals Inc.

2,2,2-trifluoro-N-(2-oxocyclohexyl)acetamide

  • MDL number MFCD28133358
  • Cas Number : 1823251-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart