LIF947290499 Life Chemicals Inc.

N-[3-(methylamino)phenyl]acetamide

  • MDL number MFCD11152638
  • Cas Number : 813425-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart