LIF947290512 Life Chemicals Inc.

6-methyl-2-(2-oxopropyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD17251805
  • Cas Number : 412018-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart