LIF947290518 Life Chemicals Inc.

2-fluoro-N,N-dimethyl-5-sulfamoylbenzamide

  • MDL number MFCD21225495
  • Cas Number : 1496857-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart