LIF947290519 Life Chemicals Inc.

3-amino-1-(furan-2-yl)propan-1-ol

  • MDL number MFCD11581655
  • Cas Number : 1082554-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart