LIF947290521 Life Chemicals Inc.

2-hydrazinyl-4,6-dimethoxypyrimidine

  • MDL number MFCD09260310
  • Cas Number : 13223-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart