LIF947290525 Life Chemicals Inc.

3-(4-fluorophenyl)prop-2-ynenitrile

  • MDL number MFCD25964863
  • Cas Number : 575433-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart