LIF947290530 Life Chemicals Inc.

6-oxo-1,6-dihydropyridine-2-carboxamide

  • MDL number MFCD11506698
  • Cas Number : 89640-67-5
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart