LIF947290534 Life Chemicals Inc.

3-benzylazetidine hydrochloride

  • MDL number MFCD16619075
  • Cas Number : 7606-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart