LIF947290538 Life Chemicals Inc.

methyl 3-aminooxolane-3-carboxylate hydrochloride

  • MDL number MFCD22369830
  • Cas Number : 1384431-37-1
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart