LIF947290539 Life Chemicals Inc.

2-{[(oxan-4-yl)methyl]amino}cyclohexan-1-ol

  • MDL number MFCD16745729
  • Cas Number : 1339678-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart