LIF947290551 Life Chemicals Inc.

3,3-difluorocyclopentane-1-carboxylic acid

  • MDL number MFCD15526661
  • Cas Number : 1260897-05-9
  • Molecular Weight : 150.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart