LIF947290572 Life Chemicals Inc.

4-(2-thienyl)piperidine-2,6-dione

  • MDL number MFCD18277040
  • Cas Number : 1267704-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart