LIF947290575 Life Chemicals Inc.

1-(5-(trifluoromethyl)pyridin-2-yl)pyrrolidin-3-ol

  • MDL number MFCD17248751
  • Cas Number : 1343181-22-5
  • Molecular Weight : 232.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart