LIF947290579 Life Chemicals Inc.

3-(pyrrolidin-3-yl)imidazolidine-2,4-dione hydrochloride

  • MDL number MFCD28017020
  • Cas Number : 1864064-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart