LIF947290581 Life Chemicals Inc.

[(1-isonicotinoylpiperidin-4-yl)methyl]amine

  • MDL number MFCD16110267
  • Cas Number : 1247536-10-2
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart